Chromate ion chromatography standard, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-ICRO-100
PROOVHG-ICRO-100EA 0 NOK
PROOVHG-ICRO-100
Chromate ion chromatography standard, VHG
Standarder Anionstandarder Chromate(VI) Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Sertifiseringer: Traceable to NIST and SRM.