Iridium standard solution, for ICP, in 20% HCl, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-PIRH-100 VHG-PIRH-500 VHG-TIRH-100 VHG-TIRH-500
PROOVHG-PIRH-100EA 0 NOK
PROOVHG-PIRH-100 PROOVHG-PIRH-500 PROOVHG-TIRH-100 PROOVHG-TIRH-500
Iridium standard solution, for ICP, in 20% HCl, VHG
Standarder Kationstandarder Iridium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifiseringer: Traceable to NIST, SRM.