Multi-element standards, major cation mix, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-SM16-500
PROOVHG-SM16-500EA 0 NOK
PROOVHG-SM16-500
Multi-element standards, major cation mix, VHG
Standarder Kationstandarder Multi-Element Standards
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Sertifiseringer: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.