Multi-element standards, comprehensive mix A, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-SM80C-500
PROOVHG-SM80C-500EA 0 NOK
PROOVHG-SM80C-500
Multi-element standards, comprehensive mix A, VHG
Standarder Kationstandarder Multi-Element Standards
This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi element standards for Ag, Al, As, Ba, Bi, Ca, Cd, Ce, Dy, Er, Eu, Ga, Gd, Ho, La, Lu, Mg, Na, Nd, P, Pb, Pr, Rb, Sc, Se, Sm, Sr, Tb, Th, Tl, Tm, U, Y, Yb at 10 µg/ml in 40% aqua regia.
Sertifiseringer: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.