Chromium standard solution, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-SPCR3-100 VHG-SPCR6-100 VHG-SPCR3-50 VHG-SPCR6-50
PROOVHG-SPCR3-100EA 0 NOK
PROOVHG-SPCR3-100 PROOVHG-SPCR6-100 PROOVHG-SPCR3-50 PROOVHG-SPCR6-50
Chromium standard solution, VHG
Standarder Kationstandarder Chromium Standards
This standard solution is intended for use as a calibration and reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.