Phosphorus standard solution, for ICP, in 5% HNO₃, VHG
Leverandør: LGC Standards PROMOCHEM
VHG-PPN-100 VHG-PPN-500 VHG-TPN-100 VHG-TPN-500
PROOVHG-PPN-100EA 0 NOK
PROOVHG-PPN-100 PROOVHG-PPN-500 PROOVHG-TPN-100 PROOVHG-TPN-500
Phosphorus standard solution, for ICP, in 5% HNO₃, VHG
Standarder Kationstandarder Phosphorus Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifiseringer: Traceable to NIST, SRM.