Anti-RG-7221 Recombinant Antibody

Leverandør: ProSci Inc.
10-664
PRSI10-664EA 8050 NOK
PRSI10-664
Anti-RG-7221 Recombinant Antibody
Antistoffer
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: V-kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR