Anti-RG-7116 Recombinant Antibody

Leverandør: ProSci Inc.
10-666
PRSI10-666EA 8050 NOK
PRSI10-666
Anti-RG-7116 Recombinant Antibody
Antistoffer
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen synonymer RO5509554|RG7155
Antistofftype Primary
Klonalitet Recombinant
Konjugering Unconjugated
Reaktivitet
Vert
Isotype IgG1 kappa
Modifikasjon None
Immunogen Humanised/CSF1R/CD115 [Homo sapiens]
Rensing >95%
Konsentrasjon Batch dependent
Lagringsbuffer PBS buffer pH 7,5
Lagringstemp Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR