Anti-AT-001 Recombinant Antibody

Leverandør: ProSci Inc.
10-820
PRSI10-820EA 8050 NOK
PRSI10-820
Anti-AT-001 Recombinant Antibody
Antistoffer
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR