Anti-Jak3 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
13-272
PRSI13-272EA 5150 NOK
PRSI13-272
Anti-Jak3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Jak3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR