Anti-RGS6 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
25-179
PRSI25-179EA 8125 NOK
PRSI25-179
Anti-RGS6 Rabbit Polyclonal Antibody
Antistoffer
Members of the RGS (regulator of G protein signaling) family, such as RGS6, modulate G protein function by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits.Members of the RGS (regulator of G protein signaling) family have been shown to modulate the functioning of G proteins by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits.

RGS6 antibody can be used for detection of RGS6 by ELISA at 1:12500. RGS6 antibody can be used for detection of RGS6 by western blot at 1 ?g/mL, and HRP conjugated secondaryMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS6
Environmentally Preferable
Antigen-navn Regulator of G-protein Signaling 6
Antigen synonymer MGC142132|DKFZp313G1241|FLJ43552|GAP
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human RGS6.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR