Anti-PAOX Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
26-838
PRSI26-838EA 8125 NOK
PRSI26-838
Anti-PAOX Rabbit Polyclonal Antibody
Antistoffer
PAOX belongs to the flavin monoamine oxidase family. PAOX is a flavoenzyme which catalyzes the oxidation of N (1)-acetylspermine to spermidine and is thus involved in the polyamine back-conversion. It can also oxidize N (1)-acetylspermidine to putrescine. PAOX does not oxidize spermidine. It plays an important role in the regulation of polyamine intracellular concentration and has the potential to act as a determinant of cellular sensitivity to the antitumor polyamine analogs.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2%More Product Information