Anti-ACO2 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
27-083
PRSI27-083EA 8125 NOK
PRSI27-083
Anti-ACO2 Rabbit Polyclonal Antibody
ACO2 belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15 (PRSS15), also known as Lon protease, after oxidative modification.The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate toMore Product Information