Anti-RGS5 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
27-222
PRSI27-222EA 8125 NOK
PRSI27-222
Anti-RGS5 Rabbit Polyclonal Antibody
Antistoffer
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators.The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are presentMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS5
Antigen-navn Regulator of G-protein Signaling 5
Environmentally Preferable
Antigen synonymer MSTP092|MSTP032|MSTP129|MST106|MSTP106|MST092|MST129
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human RGS5.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR