Anti-RGS4 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
27-303
PRSI27-303EA 8125 NOK
PRSI27-303
Anti-RGS4 Rabbit Polyclonal Antibody
Antistoffer
Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. This protein negatively regulates signaling upstream or at the level of the heterotrimeric G protein and is localizedMore Product Information