Anti-GLI2 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
27-430
PRSI27-430EA 7400 NOK
PRSI27-430
Anti-GLI2 Rabbit Polyclonal Antibody
Antistoffer
GLI2 encodes a protein which belongs to the C2H2-type zinc finger protein subclass of the Gli family. Members of this subclass are characterized as transcription factors which bind DNA through zinc finger motifs. These motifs contain conserved H-C links. Gli family zinc finger proteins are mediators of Sonic hedgehog (Shh) signaling and they are implicated as potent oncogenes in the embryonal carcinoma cell. The protein encoded by this gene localizes to the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. TheMore Product Information