Anti-RGS20 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
27-993
PRSI27-993EA 7400 NOK
PRSI27-993
Anti-RGS20 Rabbit Polyclonal Antibody
Antistoffer
Regulator of G protein signaling (RGS) proteins are regulatory and structural components of G protein-coupled receptor complexes. RGS proteins are GTPase-activating proteins for Gi and Gq class G-alpha proteins. They accelerate transit through the cycle of GTP binding and hydrolysis and thereby accelerate signaling kinetics and termination.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-More Product Information