Anti-TEF Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
28-837
PRSI28-837EA 8125 NOK
PRSI28-837
Anti-TEF Rabbit Polyclonal Antibody
Antistoffer
TEF (thyrotroph embryonic factor) is a member of the PAR bZip (proline and acidic amino acid-rich basic leucine zipper) transcription factor family. It accumulates with robust circadian rhythms in tissues with high amplitudes of clock gene expression.Thyrotroph embryonic factor (TEF), a transcription factor, is a member of the PAR (proline and acidic amino acid-rich) subfamily of basic region/leucine zipper (bZIP) transcription factors. It is expressed in a broad range of cells and tissues in adult animals, however, during embryonic development, TEF expression appears to be restricted to theMore Product Information