Anti-HSD17B6 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
29-593
PRSI29-593EA 7400 NOK
PRSI29-593
Anti-HSD17B6 Rabbit Polyclonal Antibody
Antistoffer
HSD17B6 has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convert androsterone to epi-androsterone. Both reactions use NAD+ as the preferred cofactor. HSD17B6 is a member of the retinol dehydrogenase family.The protein encoded by this gene has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convertMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HSD17B6
Environmentally Preferable
Antigen-navn Hydroxysteroid (17-beta) Dehydrogenase 6
Antigen synonymer SDR9C6|HSE|RODH
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Dog,Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunogen A synthetic peptide corresponding a region of human HSD17B6.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR