Anti-SQLE Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
29-697
PRSI29-697EA 8125 NOK
PRSI29-697
Anti-SQLE Rabbit Polyclonal Antibody
Antistoffer
Squalene epoxidase catalyzes the first oxygenation step in sterol biosynthesis and is thought to be one of the rate-limiting enzymes in this pathway.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SQLE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol SQLE
Antigen-navn Squalene Epoxidase
Environmentally Preferable
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunogen A synthetic peptide corresponding a region of human SQLE.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR