Anti-PPAP2A Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
29-703
PRSI29-703EA 7400 NOK
PRSI29-703
Anti-PPAP2A Rabbit Polyclonal Antibody
Antistoffer
PPAP2A is a member of the phosphatidic acid phosphatase (PAP) family. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. This protein is an integral membrane glycoprotein, and has been shown to be a surface enzyme that plays an active role in the hydrolysis and uptake of lipids from extracellular space. The expression of PPAP2A is found to be regulated by androgen in a prostatic adenocarcinoma cell line.The protein encoded by this gene is a member of theMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol PPAP2A
Antigen-navn Phosphatidic Acid Phosphatase Type 2A
Environmentally Preferable
Antigen synonymer PAP-2a|PAP2|LPP1|LLP1a
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Zebrafish,Human,Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human PPAP2A.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR