Anti-MTMR1 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
29-705
PRSI29-705EA 7400 NOK
PRSI29-705
Anti-MTMR1 Rabbit Polyclonal Antibody
Antistoffer
MTMR1 is a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases.This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL ofMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol MTMR1
Environmentally Preferable
Antigen-navn Myotubularin Related Protein 1
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human MTMR1.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR