Anti-RGS8 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
29-804
PRSI29-804EA 7400 NOK
PRSI29-804
Anti-RGS8 Rabbit Polyclonal Antibody
Antistoffer
RGS8 is a member of the regulator of G protein signaling (RGS) family and is a protein with a single RGS domain. Regulator of G protein signaling (RGS) proteins are regulatory and structural components of G protein-coupled receptor complexes. They accelerate transit through the cycle of GTP binding and hydrolysis to GDP, thereby terminating signal transduction, but paradoxically, also accelerate receptor-stimulated activation.This gene is a member of the regulator of G protein signaling (RGS) family and encodes a protein with a single RGS domain. Regulator of G protein signaling (RGS)More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS8
Antigen-navn Regulator of G-protein Signaling 8
Environmentally Preferable
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Dog,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human RGS8.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR