Anti-CDK9 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
31-028
PRSI31-028EA 7400 NOK
PRSI31-028
Anti-CDK9 Rabbit Polyclonal Antibody
Antistoffer
CDK9 is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and known as important cell cycle regulators. This kinase was found to be a component of the multiprotein complex TAK/P-TEFb, which is an elongation factor for RNA polymerase II-directed transcription and functions by phosphorylating the C-terminal domain of the largest subunit of RNA polymerase II. This protein forms a complex with and is regulated by its regulatory subunit cyclin T or cyclin K. HIV-1 Tat protein wasMore Product Information