Anti-RGS10 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
31-069
PRSI31-069EA 7400 NOK
PRSI31-069
Anti-RGS10 Rabbit Polyclonal Antibody
Antistoffer
Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 10 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. This protein associates specifically with the activated forms of the two related G-protein subunits, G-alphai3 and G-More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS10
Antigen-navn Regulator of G-protein Signaling 10
Environmentally Preferable
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human RGS10.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR