Anti-GABRA3 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
31-103
PRSI31-103EA 7400 NOK
PRSI31-103
Anti-GABRA3 Rabbit Polyclonal Antibody
Antistoffer
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GABRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol GABRA3
Environmentally Preferable
Antigen-navn Gamma-aminobutyric acid (GABA) A Receptor alpha 3
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human GABRA3.
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR