Anti-GABRP Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
31-108
PRSI31-108EA 7400 NOK
PRSI31-108
Anti-GABRP Rabbit Polyclonal Antibody
Antistoffer
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. The subunit encoded by this gene is expressed in several non-neuronal tissues including the uterus and ovaries. This subunit can assemble with known GABA A receptor subunits, and the presence of this subunit alters the sensitivity of recombinant receptors to modulatory agents such as pregnanolone.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.More Product Information