Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: HS402]

Leverandør: ProSci Inc.
36-135
PRSI36-135EA 8450 NOK
PRSI36-135
Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: HS402]
Antistoffer
TRAIL-R4 is a receptor for the cytotoxic ligand TRAIL. Contains a truncated death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.

Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: HS402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol TNFRSF10D
Environmentally Preferable
Antigen-navn Tumor Necrosis Factor Receptor Superfamily Member 10D
Antigen synonymer TRAILR4|TRAIL-R4|CD264|TRUNDD|DCR2
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon HS402
Reaktivitet Human
Vert Mouse
Gen-ID 8793
Isotype IgG1
ImmunoKjemi Yes
Immunutfelling Yes
Flowcytometri Yes
Immunogen Recombinant human TRAIL-R4 (DcR2).
Skjema Liquid
Rensing >95% (SDS-PAGE)
Konsentrasjon 1 mg/ml
Lagringsbuffer In PBS containing 10% glycerol and 0.02% sodium azide.
Lagringstemp Stable for at least 1 year after receipt when stored at -20˚C.
Frakttemp. -20°C
Uttestet applikasjon FACS,

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR