Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]

Leverandør: ProSci Inc.
36-148
PRSI36-148EA 5775 NOK
PRSI36-148
Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]
Antistoffer
CD40 belongs to the TNF-receptor superfamily and is essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. The interaction of CD40-CD40L is necessary for amyloid-beta-induced microglial activation, and thus is thought to be an early event in Alzheimer disease pathogenesis. CD40 is constitutively expressed by antigen presenting cells, including dendritic cells, B cells and macrophages. Consistent with its widespread expression on normal cells, CD40 is alsoMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CD40
Antigen-navn CD40 antigen
Environmentally Preferable
Antigen synonymer CD40L Receptor|Tumor Necrosis Factor Receptor Superfamily Member 5
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon FGK45
Reaktivitet Mouse
Vert Rat
Gen-ID 21939
Isotype IgG2a
Flowcytometri Yes
Immunogen Recombinant mouse CD40 fusion protein.
Skjema Liquid
Rensing >95% (SDS-PAGE)
Konsentrasjon 1 mg/ml
Lagringsbuffer In PBS. Preservative free formulation.
Lagringstemp Stable for at least 1 year after receipt when stored at -20˚C.
Frakttemp. -20°C
Uttestet applikasjon FACS, FUNC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR