Anti-CFLAR Mouse Monoclonal Antibody (Biotin) [clone: NF6]

Leverandør: ProSci Inc.
36-163
PRSI36-163EA 7225 NOK
PRSI36-163
Anti-CFLAR Mouse Monoclonal Antibody (Biotin) [clone: NF6]
Antistoffer
FLIP is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells and acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. FLIP lacks enzymatic (caspase) activity. FLIP is highly expressed in skeletal muscle, pancreas, heart, kidney,More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CFLAR
Environmentally Preferable
Antigen-navn CASP8 and FADD-like apoptosis regulator
Antigen synonymer CASPER|CLARP|FLAME-1|I-FLICE|Usurpin|CASH
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Biotin
Klon NF6
Reaktivitet Human
Vert Mouse
Gen-ID 8837
Isotype IgG1
Western blot Yes
ImmunoKjemi Yes
Immunogen Recombinant human FLIP (aa 1-480).
Skjema Liquid
Konsentrasjon 1 mg/ml
Lagringsbuffer In PBS containing 10% glycerol and 0.02% sodium azide.
Lagringstemp Stable for at least 1 year after receipt when stored at -20˚C.
Frakttemp. -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR