Anti-GRN Guinea Pig Polyclonal Antibody
Leverandør: ProSci Inc.
36-314
PRSI36-314EA 6975 NOK
PRSI36-314
Anti-GRN Guinea Pig Polyclonal Antibody
Antistoffer
Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease (AD) and potentially for MCI (Mild Cognitive Impairment). Additionally, PGRN isMore Product Information