Anti-GRTP1 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
4591
PRSI4591EA 5350 NOK
PRSI4591
Anti-GRTP1 Rabbit Polyclonal Antibody
Antistoffer
GRTP1 Antibody: Growth hormone regulated TBC protein 1 (GRTP1) was identified through microarray analysis as a gene whose expression is regulated by growth hormone (GH). It contains the TBC signature motif of GTPase activator proteins of Rab-like small GTPases. GRTP1 mRNA is expressed at highest levels in testes, and is also present in kidney, lung, liver, and intestine. Administration of GH to mice resulted in an increase in GRTP1 mRNA in testes, but a decrease of GRTP1 mRNA in kidney and liver. GRTP1 was identified as an HIV dependency factor (HDF), suggesting that GRTP1 may be anMore Product Information