Anti-GSK3B Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
49-518
PRSI49-518EA 10025 NOK
PRSI49-518
Anti-GSK3B Rabbit Polyclonal Antibody
Antistoffer
Glycogen synthase kinase-3 (GSK 3) is a proline-directed serine-threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase. Two isoforms, alpha (GSK 3A) and beta, show a high degree of amino acid homology. GSK 3B is involved in energy metabolism, neuronal cell development, and body pattern formation. GSK 3B participates in the Wnt signaling pathway and has been implicated in the hormonal control of several regulatory proteins including glycogen synthase, MYB and the transcription factor JUN. GSK 3B phosphorylates JUN at sites proximal to its DNA-More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GSK3B
Antigen-navn Glycogen Synthase Kinase 3 beta
Environmentally Preferable
Antigen synonymer EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Immunogen GSK3B antibody was raised against phospho amino acids 4 - 12 of GSK3B (Human).
Skjema Liquid
Uttestet applikasjon Immunoaffinity Chromatography

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR