Anti-CERS6 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
4941
PRSI4941EA 5350 NOK
PRSI4941
Anti-CERS6 Rabbit Polyclonal Antibody
Antistoffer
LASS6 Antibody: The LASS (longevity assurance homolog) family members represent a subgroup of the homeobox gene family and are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1-6) and all are involved in ceramide synthesis during cell growth regulation and cancer differentiation. Like the highly homologous LASS5, LASS6 is also an endoplasmic reticulum, multi-pass membrane protein. LASS6 is also involved in the synthesis of C14, C16 and C18-ceramide, but shows a preference for unsaturated fatty acids. LASS6 is broadly expressed in aMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CERS6
Antigen-navn Ceramide Synthase 6
Environmentally Preferable
Antigen synonymer CerS6|MGC129949|MGC129950
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen LASS6 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of the human LASS6.
Rensing Affinity chromatography purified via peptide column
Lagringsbuffer PBS containing 0.02% sodium azide
Uttestet applikasjon E, WB, IHC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR