Anti-RGS21 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
5209
PRSI5209EA 5350 NOK
PRSI5209
Anti-RGS21 Rabbit Polyclonal Antibody
Antistoffer
RGS21 Antibody: Regulator of G-protein signaling (RGS) proteins contain an 120 amino acid conserved domain, termed the RGS domain, that acts as a GTPase-activating protein that acts to reduce the signal transmitted by the receptor-activated G-alpha subunit. RGS21 is a recently identified member of this family that has been reported to be selectively expressed in subpopulations of taste bud cells and co-expressed with bitter and sweet transduction components such as alpha-gusticin, phospholipase Cbeta2, T1R2/T1R3 sweet taste receptors and T2R bitter taste receptors. Other reports indicateMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS21
Antigen-navn Regulator of G-protein Signaling 21
Environmentally Preferable
Antigen synonymer Regulator of G-protein signaling 21|RGS21|RGS21_HUMAN.
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen RGS21 antibody was raised against a 16 amino acid synthetic peptide near the center of human RGS21.
Rensing Affinity chromatography purified via peptide column
Lagringsbuffer PBS containing 0.02% sodium azide
Uttestet applikasjon E, WB, IHC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR