Anti-PAPPA Monoclonal Antibody [clone: A1]

Leverandør: ProSci Inc.
54-308
PRSI54-308EA 8050 NOK
PRSI54-308
Anti-PAPPA Monoclonal Antibody [clone: A1]
Antistoffer
Type: Primary
Antigen: PAPPA
Clonality: Monoclonal
Clone: A1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR