Anti-PAPPA Monoclonal Antibody [clone: C1]

Leverandør: ProSci Inc.
54-309
PRSI54-309EA 8050 NOK
PRSI54-309
Anti-PAPPA Monoclonal Antibody [clone: C1]
Antistoffer
Type: Primary
Antigen: PAPPA
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR