Anti-PLEKHM3 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
5487
PRSI5487EA 5350 NOK
PRSI5487
Anti-PLEKHM3 Rabbit Polyclonal Antibody
Antistoffer
PLEKHM3 Antibody: PLEKHM3, also known as DAPR, is a member of the M family of Pleckstrin homology domain-containing proteins. PLEKHM3 was initially identified through chromatin immunoprecipitation and CpG microarray analysis examining proteins regulated by myocyte-enhancing factor 2. In C2C12 myoblast cells, PLEKHM3 binds to the PI3K signaling member protein kinase B in the cytosol prior to differentiation into myotubes. Following the initiation of differentiation, PLEKHM3 was also found in membrane fractions. Knockdown of PLEKHM3 expression by RNAi resulted in the inhibition of myotubeMore Product Information