Anti-HMOX1 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
55-728
PRSI55-728EA 7625 NOK
PRSI55-728
Anti-HMOX1 Rabbit Polyclonal Antibody
Antistoffer
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.

For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR