Anti-SNX13 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
56-298
PRSI56-298EA 7625 NOK
PRSI56-298
Anti-SNX13 Rabbit Polyclonal Antibody
Antistoffer
This gene encodes a PHOX domain- and RGS domain-containing protein that belongs to the sorting nexin (SNX) family and the regulator of G protein signaling (RGS) family. The PHOX domain is a phosphoinositide binding domain, and the SNX family members are involved in intracellular trafficking. The RGS family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. The RGS domain of this protein interacts with G alpha(s), accelerates its GTP hydrolysis, and attenuates G alpha(s)-mediated signaling. Overexpression of this proteinMore Product Information