Anti-PDE6A Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
56-928
PRSI56-928EA 7625 NOK
PRSI56-928
Anti-PDE6A Rabbit Polyclonal Antibody
Antistoffer
This gene encodes the cyclic-GMP (cGMP)-specific phosphodiesterase 6A alpha subunit, expressed in cells of the retinal rod outer segment. The phosphodiesterase 6 holoenzyme is a heterotrimer composed of an alpha, beta, and two gamma subunits. cGMP is an important regulator of rod cell membrane current, and its dynamic concentration is established by phosphodiesterase 6A cGMP hydrolysis and guanylate cyclase cGMP synthesis. The protein is a subunit of a key phototransduction enzyme and participates in processes of transmission and amplification of the visual signal. Mutations in this geneMore Product Information