Anti-TMEM184C Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
5685
PRSI5685EA 5350 NOK
PRSI5685
Anti-TMEM184C Rabbit Polyclonal Antibody
Antistoffer
TMEM184C Antibody: Anaplastic thyroid cancer is one of the most lethal forms of cancer, but the precise carcinogenic mechanism has not been identified. TMEM184C, also known as TMEM34, was identified in a cDNA microarray analysis as being down-regulated in anaplastic thyroid cancers compared to normal thyroid tissues. TMEM184C protein expression was also lower in cell lines derived from these types of cancers compared to that of normal thyroid tissues or cell lines based on other types of thyroid cancers. Furthermore, transfection of TMEM34 into KTA2 cells led to the inhibition of cell growth,More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol TMEM184C
Antigen-navn Transmembrane Protein 184C
Environmentally Preferable
Antigen synonymer TMEM34|Transmembrane protein 184C|TMEM184C|PRO1355|Transmembrane protein 34
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen TMEM184C antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human TMEM184C.
Rensing Affinity chromatography purified via peptide column
Lagringsbuffer PBS containing 0.02% sodium azide
Uttestet applikasjon E, WB, IHC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR