Anti-AFP Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
5869
PRSI5869EA 5350 NOK
PRSI5869
Anti-AFP Rabbit Polyclonal Antibody
Antistoffer
AFP Antibody: Alpha-fetoprotein (AFP) is a major plasma protein produced by the yolk sac and the liver during fetal development. In adults, AFP expression is often associated with hepatoma or teratoma. However, hereditary persistance of AFP may also be found in individuals with no obvious pathology. AFP is thought to be the fetal counterpart of serum albumin, and the AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. AFPMore Product Information