Anti-ZFP281 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
6233
PRSI6233EA 5350 NOK
PRSI6233
Anti-ZFP281 Rabbit Polyclonal Antibody
Antistoffer
ZFP281 Antibody: ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, andMore Product Information