Anti-CDCP1 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
79-675
PRSI79-675EA 5675 NOK
PRSI79-675
Anti-CDCP1 Rabbit Polyclonal Antibody
Antistoffer
May be involved in cell adhesion and cell matrix association. May play a role in the regulation of anchorage versus migration or proliferation versus differentiation via its phosphorylation. May be a novel marker for leukemia diagnosis and for immature hematopoietic stem cell subsets. Belongs to the tetraspanin web involved in tumor progression and metastasis.
Western Blot: 1:500
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50%More Product Information