Human recombinant NAPRTase (from E. coli)
Leverandør: ProSci Inc.
90-363
PRSI90-363EA 9500 NOK
PRSI90-363
Human recombinant NAPRTase (from E. coli)
Enzymer
Human nicotinate phosphoribosyltransferase (NAPRTase) is localised in the cytoplasm and is involved in biological processes such as NAD biosynthetic and metabolic processes, nicotinamide metabolic process, nicotinate nucleotide salvage, response to oxidative stress and water-soluble vitamin metabolic process. It functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN) and is essential for NA to increase cellular NAD levels and prevent oxidative stress of the cells. It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.