Mouse recombinant IL2 (from CHO cells)
Leverandør: ProSci Inc.
90-467
PRSI90-467EA 7225 NOK
PRSI90-467
Mouse recombinant IL2 (from CHO cells)
Proteiner
Interleukin-2 (IL-2) is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells, and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils, and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta. Moreover, studies have shown that IL-2 is required for activation-induced apoptosis, an important hemeostatic mechanism in the immune system, which is involved in the maintenance of peripheral tolerance to self-antigens.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.