Human Recombinant DHS (from E. coli)
Leverandør: ProSci Inc.
91-112
PRSI91-112EA 8050 NOK
PRSI91-112
Human Recombinant DHS (from E. coli)
Proteiner
Human Deoxyhypusine Synthase (DHS) is vital for the first step of hypusine biosynthesis. DHS catalyzes the NAD-dependent oxidative cleavage of spermidine, the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag