Human recombinant FABP6 (from E. coli)

Leverandør: ProSci Inc.

91-201
PRSI91-201EA 10875 NOK
PRSI91-201
Human recombinant FABP6 (from E. coli)
Proteiner

Fatty Acid-Binding Protein 6 (FABP6) is cytoplasmic protein that binds long-chain fatty acids and other hydrophobic ligands which belongs to the calycin superfamily. FABP6 expression is restricted in the small intestine to the ileum where it is involved in the enterohepatic circulation of bile acids. FABP6 forms a beta-barrel structure that accommodates the hydrophobic ligand in its interior. Isoform 2 is expressed in colorectal adenocarcinomas and their adjacent normal mucosa (at protein level). Isoform 1 is expressed in the jejunum, ileum, cecum and ascending colon intestine. FABP6 plays a role in fatty acid uptake, transport, and metabolism. FABP6 stimulates gastric acid and pepsinogen secretion. It seems to be able to bind to bile salts and bilirubins.


  • This recombinant protein can be used for biological assays. For research use only.


Fusion-Tag: N-6 His tag

Bestill nå

SPESIFIKASJONER

Protein/peptide name FABP6
Environmentally Preferable
Protein synonyms Fatty Acid-Binding Protein 6|Intestinal Bile Acid-Binding Protein|FABP6|I-BABP|ILLBP|Gastrotropin|ILBP|I-15P|Ileal Lipid-Binding Protein|Intestinal 15 kDa Protein|GT
Protein/peptide type Recombinant
Arter Human
Kilde E. coli
Sekvens Met1-Ala128
Renhet > 95% as determined by reducing SDS-PAGE
Formulering Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 0.5mM DTT, 50% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Molekylvekt 16.6 kD
Lagringsforhold Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
Tag sekvens N-6 His tag
Uttestet applikasjon For Biological assays

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR