Human recombinant ULBP-2 (from cells)

Leverandør: ProSci Inc.

91-459
PRSI91-459EA 6500 NOK
PRSI91-459
Human recombinant ULBP-2 (from cells)
Proteiner

NKG2D Ligand 2 (N2DL2) is a member of a family of cell-surface proteins. N2DL2 function as ligands for human cytomegalovirus glycoprotein UL16. N2DL2 is anchored to the membrane via a GPI-linkage. N2DL2 is bind to human NKG2D, an activating receptor expressed on NK cells, NKT cells, T cells. Engagement of NKG2D results in the activation of cytolytic activity and cytokine production by these effects cells. The ULBPs are expressed on some tumor cells and have been implicated in tumor surveillance.


  • This recombinant protein can be used for biological assays. For research use only.


Fusion-Tag: C-6 His tag

Bestill nå

SPESIFIKASJONER

Protein/peptide name ULBP-2
Environmentally Preferable
Protein synonyms NKG2DL2|ULBP2|UL16-Binding Protein 2|N2DL2|NKG2D Ligand 2|N2DL-2|Retinoic Acid Early Transcript 1H|ALCAN-Alpha|RAET1H
Protein/peptide type Recombinant
Arter Human
Kilde Cells
Sekvens Gly26-Ser217
Renhet > 95% as determined by reducing SDS-PAGE
Formulering Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Molekylvekt 22.78 kD
Lagringsforhold Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tag sekvens C-6 His tag
Uttestet applikasjon For Biological Assays

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR